Learn More
Enzo Life Sciences GroES (E. coli), (recombinant) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPP620F
Encompass_Preferred
Alternative name: Heat shock protein 10, HSP10, 10kDa Chaperonin, Protein Cpn10 GroES protein. Purity: ≥95% (SDS-PAGE; Western blot). MW: ~10kDa. Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 100mM sodium chloride, 10mM magnesium chloride, and 5mM DTT. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7611