Learn More
Enzo Life Sciences GroES (E. coli), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISPP620D
Encompass_Preferred
Alternative name: Heat shock protein 10, HSP10, 10kDa Chaperonin, Protein Cpn10 GroES protein. MW: ~10kDa. Purity: ≥95% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 100mM sodium chloride, 10mM magnesium chloride, and 5mM DTT. Source: Produced in E. coli. UniProt: P0A6F9 (strain K12). Application: WB. Long Term Storage: -80°C
Catalog No. 50-950-082