Learn More
Advanced Medical Systems GroMed™ 5 units/case
Catalog No. NC0943529
Encompass_Preferred
Personal Aseptic Technique Test (PATT2) kit. Ideal for facilities manipulating ampules. Easy to use kit for testing up to 5 individuals. 20ml vials of TSB media.100ml partially filled minibags for added complexity - 3ml ampules of colored TSB media -Log sheet included -Gummed labels included -Product conforms to United States Pharmacopeia standards
Catalog No. NC0943529 Supplier Advanced Medical Systems Supplier No. PMTBVA123
Personal Aseptic Technique Test (PATT2) kit. Ideal for facilities manipulating ampules. Easy to use kit for testing up to 5 individuals. 20ml vials of TSB media.100ml partially filled minibags for added complexity - 3ml ampules of colored TSB media -Log sheet included -Gummed labels included -Product conforms to United States Pharmacopeia standards