Learn More
Grainger Grommet for TV-416 .453 PK25
Catalog No. 502750918
Encompass_Preferred
The Haltec 140V12 is a rubber grommet with an inside diameter of 1/2", an outside diameter of 3/4", and a panel thickness capacity of 1/8" to 1/4". It is made of EPDM rubber and has a temperature range of -40°F to 250°F. The grommets have a tensile strength of 1500 psi and 500% elongation. They come in a package of 100 pieces and are RoHS compliant. The grommets are designed for use in electronic enclosures and provide insulation and strain relief for wires and cables. Their flexible rubber construction allows wires and cables to pass through smoothly while sealing out dust, moisture and noise.
Catalog No. 50-275-0918 Supplier Grainger Supplier No. 140V12
The Haltec 140V12 is a rubber grommet with an inside diameter of 1/2", an outside diameter of 3/4", and a panel thickness capacity of 1/8" to 1/4". It is made of EPDM rubber and has a temperature range of -40°F to 250°F. The grommets have a tensile strength of 1500 psi and 500% elongation. They come in a package of 100 pieces and are RoHS compliant. The grommets are designed for use in electronic enclosures and provide insulation and strain relief for wires and cables. Their flexible rubber construction allows wires and cables to pass through smoothly while sealing out dust, moisture and noise.