Learn More
MSC Value Collection Grommet, 7/8 in x 1-5/8 in, Rubber, 25/Pack
Catalog No. 502866037
Encompass
This rubber grommet features an inside diameter of 7/8 in, an outside diameter of 1-5/8 in, and an overall thickness of 7/16 in. It is designed for various applications requiring insulation or protection for openings.
  • Material: rubber
  • Inside diameter: 7/8 in
  • Slot diameter: 1-1/4 in
  • Slot width: 1/16 in
  • Overall thickness: 7/16 in
  • Outside diameter: 1-5/8 in
Catalog No. 50-286-6037 Supplier MSC Supplier No. 66453069
This rubber grommet features an inside diameter of 7/8 in, an outside diameter of 1-5/8 in, and an overall thickness of 7/16 in. It is designed for various applications requiring insulation or protection for openings.
  • Material: rubber
  • Inside diameter: 7/8 in
  • Slot diameter: 1-1/4 in
  • Slot width: 1/16 in
  • Overall thickness: 7/16 in
  • Outside diameter: 1-5/8 in