Learn More
Ampco Groove Chisel with Flat Tip
SDP
Catalog No. 19173539 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
10 -1/4 in.
Catalog No. Length (English)
19-173-539 10 -1/4 in.
1 options
Catalog No. 19-173-539 Supplier Ampco Supplier No. 41035
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant groove chisel. Made in Germany.
  • See image for additional information regarding measurements.
  • Tip height (a): 5/16 in.
  • Tip width (b): 5/8 in.
  • Overall length: 10 and 1/4 in.
  • Shaft diameter (s): 1/2 in.
Specifications
Product Type Caulking Tool
Length (English) 10 -1/4 in.
Quantity 1 Ea.