Learn More
Grainger Groove Tool Retaining Ring Set 6 Pc
Catalog No. 502766803
Encompass_Preferred
The Micro 100 Grooving Tool Set is a 3-piece set of grooving tools with a 3/8" shank diameter. The set includes a 0.040" grooving tool, a 0.062" grooving tool, and a 0.125" grooving tool, allowing a range of groove widths. The tools have a carbide insert with a TiAlN coating for wear resistance and heat dissipation. They are designed for grooving, cutoff, and parting applications in steel, stainless steel, cast iron, aluminum, and other materials. The tools have a maximum depth of cut of 0.197" and can machine at up to 5000 RPM. This versatile set of Micro 100 grooving tools allows efficient grooving, cutoff, and parting operations.
Catalog No. 50-276-6803 Supplier Grainger Supplier No. 16T696
The Micro 100 Grooving Tool Set is a 3-piece set of grooving tools with a 3/8" shank diameter. The set includes a 0.040" grooving tool, a 0.062" grooving tool, and a 0.125" grooving tool, allowing a range of groove widths. The tools have a carbide insert with a TiAlN coating for wear resistance and heat dissipation. They are designed for grooving, cutoff, and parting applications in steel, stainless steel, cast iron, aluminum, and other materials. The tools have a maximum depth of cut of 0.197" and can machine at up to 5000 RPM. This versatile set of Micro 100 grooving tools allows efficient grooving, cutoff, and parting operations.