Learn More
Grainger Kyocera Indexable Parting and Grooving Insert, Carbide, PR1215, 5/Pk.
Catalog No. 502775795
Encompass_Preferred
This indexable parting and grooving insert is designed for precision external grooving applications. Constructed from durable carbide and featuring a PVD coating, the insert offers high performance and extended tool life in demanding machining environments.
  • Designed for right-hand external grooving
  • Features a chip-breaker to improve chip control
  • Constructed from carbide for enhanced wear resistance
  • Coated with PVD for superior thermal and chemical stability
  • Insert seat size of 32
  • Cutter width of 0.0945 in.
  • Radius of 0.047 in.
  • Inscribed circle of 3/8 in.
  • Insert thickness of 1/8 in.
  • Sold in multiples of 5 pieces
Catalog No. 50-277-5795 Supplier Grainger Supplier No. 170MW0
This indexable parting and grooving insert is designed for precision external grooving applications. Constructed from durable carbide and featuring a PVD coating, the insert offers high performance and extended tool life in demanding machining environments.
  • Designed for right-hand external grooving
  • Features a chip-breaker to improve chip control
  • Constructed from carbide for enhanced wear resistance
  • Coated with PVD for superior thermal and chemical stability
  • Insert seat size of 32
  • Cutter width of 0.0945 in.
  • Radius of 0.047 in.
  • Inscribed circle of 3/8 in.
  • Insert thickness of 1/8 in.
  • Sold in multiples of 5 pieces