Learn More
Grainger Grooving GDM 4020N080GM PR1535 G 10 pcs.
Catalog No. 502764201
Encompass_Preferred
The 170NF4 is a capacitor manufactured by Kemet. It is a non-polarized, film capacitor with a capacitance value of 0.1 microfarads and a tolerance of +/- 5%. The capacitor has a rated voltage of 400VDC and a maximum operating temperature range of -55°C to 125°C. The capacitor uses polypropylene film as the dielectric and has axial leads. Key features of this capacitor are its high insulation resistance, low dissipation factor, and stability over temperature. With its 400V rating, this capacitor is well-suited for general purpose filtering, coupling, and bypassing applications.
Catalog No. 50-276-4201 Supplier Grainger Supplier No. 170NF4
The 170NF4 is a capacitor manufactured by Kemet. It is a non-polarized, film capacitor with a capacitance value of 0.1 microfarads and a tolerance of +/- 5%. The capacitor has a rated voltage of 400VDC and a maximum operating temperature range of -55°C to 125°C. The capacitor uses polypropylene film as the dielectric and has axial leads. Key features of this capacitor are its high insulation resistance, low dissipation factor, and stability over temperature. With its 400V rating, this capacitor is well-suited for general purpose filtering, coupling, and bypassing applications.