Learn More
Grainger Kyocera Indexable Parting and Grooving Insert, Carbide, 5 mm Cutting Width, 2.5 mm Corner Radius, 10/Pk.
Catalog No. 502774906
Encompass_Preferred
This item has to be ordered in multiples of 10 per supplier requirements.
These indexable parting and grooving inserts are designed for efficient material removal in production turning applications, including copying and internal diameter grooving. Engineered for high-volume metalworking, these carbide inserts support high speeds and feeds while minimizing tool change downtime in lathes and CNC turning machines.
  • Carbide material with Megacoat coating and PVD coating process for enhanced durability
  • Neutral cutting direction with two cutting edges
  • Compatible with KGDIL-4, KGDIL-5, KGDIR-4, and KGDIR-5 toolholders
  • 5.00 mm (0.1969 in.) cutting width and 11.00 mm (0.4330 in.) maximum depth of cut
  • 2.50 mm (0.0984 in.) corner radius
  • Clamp-style holding method for secure mounting
Catalog No. 50-277-4906 Supplier Grainger Supplier No. 170MM0
This item has to be ordered in multiples of 10 per supplier requirements.
These indexable parting and grooving inserts are designed for efficient material removal in production turning applications, including copying and internal diameter grooving. Engineered for high-volume metalworking, these carbide inserts support high speeds and feeds while minimizing tool change downtime in lathes and CNC turning machines.
  • Carbide material with Megacoat coating and PVD coating process for enhanced durability
  • Neutral cutting direction with two cutting edges
  • Compatible with KGDIL-4, KGDIL-5, KGDIR-4, and KGDIR-5 toolholders
  • 5.00 mm (0.1969 in.) cutting width and 11.00 mm (0.4330 in.) maximum depth of cut
  • 2.50 mm (0.0984 in.) corner radius
  • Clamp-style holding method for secure mounting