Learn More
Grainger Grooving GDM 6020N040GL PR1535 G 10 pcs.
Catalog No. 502764249
Encompass_Preferred
The Grainger 170NF9 is a 9-inch, non-fused disconnect switch. It is rated for 600 volts AC and has a 100 ampere current rating. The switch has a NEMA 1 general purpose enclosure constructed from steel with a gray baked enamel finish. It features a quick-make, quick-break toggle operating mechanism with the ability to be padlocked in the OFF position. This disconnect switch can be mounted to walls or panels using the mounting holes located on the enclosure. It is UL listed and CSA certified. The 170NF9 provides a safe and reliable method for manually disconnecting and isolating electrical circuits.
Catalog No. 50-276-4249 Supplier Grainger Supplier No. 170NF9
The Grainger 170NF9 is a 9-inch, non-fused disconnect switch. It is rated for 600 volts AC and has a 100 ampere current rating. The switch has a NEMA 1 general purpose enclosure constructed from steel with a gray baked enamel finish. It features a quick-make, quick-break toggle operating mechanism with the ability to be padlocked in the OFF position. This disconnect switch can be mounted to walls or panels using the mounting holes located on the enclosure. It is UL listed and CSA certified. The 170NF9 provides a safe and reliable method for manually disconnecting and isolating electrical circuits.