Learn More
Grainger Kyocera Indexable Parting and Grooving Insert, Carbide, 6.00 mm, 2 Edges, 10/Pk.
Catalog No. 502777254
Encompass_Preferred
These indexable parting and grooving inserts are designed for efficient workpiece cutoff and grooving in turning applications. Engineered for performance in high-speed and high-feed metalworking, these carbide inserts reduce tool change downtime by allowing for quick indexing to a fresh cutting edge. They are optimized for machining steel and stainless steel in lathes and CNC turning centers.
  • Carbide material with MEGACOAT finish for enhanced durability
  • Positive rake angle of 2.000°
  • Features two cutting edges for extended tool life
  • Designed for neutral cutting direction
  • Includes GM chip breaker for effective chip control
  • Maximum depth of cut of 30.00 mm (1.1811 in.)
  • Cutting width of 6.00 mm (0.2362 in.)
Catalog No. 50-277-7254 Supplier Grainger Supplier No. 170MM6
These indexable parting and grooving inserts are designed for efficient workpiece cutoff and grooving in turning applications. Engineered for performance in high-speed and high-feed metalworking, these carbide inserts reduce tool change downtime by allowing for quick indexing to a fresh cutting edge. They are optimized for machining steel and stainless steel in lathes and CNC turning centers.
  • Carbide material with MEGACOAT finish for enhanced durability
  • Positive rake angle of 2.000°
  • Features two cutting edges for extended tool life
  • Designed for neutral cutting direction
  • Includes GM chip breaker for effective chip control
  • Maximum depth of cut of 30.00 mm (1.1811 in.)
  • Cutting width of 6.00 mm (0.2362 in.)