Learn More
Grainger Grooving GDM 6020N080GM PR1535 G 10 pcs.
Catalog No. 502764187
Encompass_Preferred
The Grainger 170NG1 is a 1/2 HP, 115/230V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be wired for 115V or 230V operation. The motor is CSA certified and meets NEMA MG1 standards. With its 1/2 HP and 1725 RPM, this electric motor provides efficient and reliable power for industrial, commercial, and residential applications.
Catalog No. 50-276-4187 Supplier Grainger Supplier No. 170NG1
The Grainger 170NG1 is a 1/2 HP, 115/230V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body, aluminum end brackets, and double sealed ball bearings. It can be wired for 115V or 230V operation. The motor is CSA certified and meets NEMA MG1 standards. With its 1/2 HP and 1725 RPM, this electric motor provides efficient and reliable power for industrial, commercial, and residential applications.