Learn More
Grainger Grooving GDM 6020N300RCM PR1535 10 pcs.
Catalog No. 502764188
Encompass_Preferred
The Grainger 170NG2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Sleeve Bearing, General Purpose Motor. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a speed of 1725 RPM, uses a 56C frame, is open drip proof, utilizes ball and sleeve bearings, and is designed for general purpose applications. The motor is compact and versatile for driving pumps, fans, conveyors, packaging equipment, and other machinery.
Catalog No. 50-276-4188 Supplier Grainger Supplier No. 170NG2
The Grainger 170NG2 is a Dayton 1/3 HP, 115/230V AC, 1725 RPM, 56C Frame, Open Drip Proof, Ball Bearing, Sleeve Bearing, General Purpose Motor. It has a 1/3 horsepower, operates on 115/230 volt AC power, has a speed of 1725 RPM, uses a 56C frame, is open drip proof, utilizes ball and sleeve bearings, and is designed for general purpose applications. The motor is compact and versatile for driving pumps, fans, conveyors, packaging equipment, and other machinery.