Learn More
Grainger Grooving KCG 3125R A65 Unc 10 pcs.
Catalog No. 502764193
Encompass_Preferred
The 170NM5 is a 5-gallon plastic pail manufactured by Encore Plastics. It has an outside diameter of 12.75 inches and a height of 15.5 inches. The pail is made from high density polyethylene and has a screw-on lid with a gasket seal. It can hold up to 60 pounds and is suited for a variety of uses including mixing, storing and transporting viscous materials. The pail is FDA-compliant and BPA-free. It provides an economical and durable solution for containing bulk liquids and materials.
Catalog No. 50-276-4193 Supplier Grainger Supplier No. 170NM5
The 170NM5 is a 5-gallon plastic pail manufactured by Encore Plastics. It has an outside diameter of 12.75 inches and a height of 15.5 inches. The pail is made from high density polyethylene and has a screw-on lid with a gasket seal. It can hold up to 60 pounds and is suited for a variety of uses including mixing, storing and transporting viscous materials. The pail is FDA-compliant and BPA-free. It provides an economical and durable solution for containing bulk liquids and materials.