Learn More
Grainger Micro 100 Grooving Tool, Solid Carbide, Right Hand, 1 ea
Catalog No. 502788185
Encompass_Preferred
This solid carbide grooving tool is designed for lathes, machining centers, and Swiss type screw machines to perform precise face or internal grooving operations, such as creating retaining ring recesses. Featuring a right-hand cutting direction, this tool is engineered to reduce setup time and improve productivity across a variety of materials including steel, stainless steel, HRSA, non-ferrous metals, cast iron, and plastics.
  • Features a unique 2-point locking and locating system ensuring tool-to-tool change repeatability of ±0.0005 in. centerline.
  • Equipped with elliptical necks for maximum rigidity during operation.
  • Compatible with a wide range of machines including Tsugami, Starr, Okuma, Omni-Turn, Citizens, Tornos, Haas, and Hardinge systems.
  • Designed for high-performance applications in industries such as medical, aerospace, automotive, and firearms manufacturing.
Catalog No. 50-278-8185 Supplier Grainger Supplier No. 16L683
This solid carbide grooving tool is designed for lathes, machining centers, and Swiss type screw machines to perform precise face or internal grooving operations, such as creating retaining ring recesses. Featuring a right-hand cutting direction, this tool is engineered to reduce setup time and improve productivity across a variety of materials including steel, stainless steel, HRSA, non-ferrous metals, cast iron, and plastics.
  • Features a unique 2-point locking and locating system ensuring tool-to-tool change repeatability of ±0.0005 in. centerline.
  • Equipped with elliptical necks for maximum rigidity during operation.
  • Compatible with a wide range of machines including Tsugami, Starr, Okuma, Omni-Turn, Citizens, Tornos, Haas, and Hardinge systems.
  • Designed for high-performance applications in industries such as medical, aerospace, automotive, and firearms manufacturing.