Learn More
Echelon Biosciences Research Labs GROPI 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs P00001MG
Encompass
Glycerophosphoinositol (GroPI) is the building block for phosphoinositide polyphosphates and is a substrate for PI-PLC. This item is non returnable
Catalog No. 50-720-542