Learn More
Echelon Biosciences Research Labs GROPI 500 ug
SDP
Catalog No. 50720544
Encompass
Glycerophosphoinositol (GroPI) is the building block for phosphoinositide polyphosphates and is a substrate for PI-PLC. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-544 Supplier Echelon Biosciences Research Labs Supplier No. P0000500UG
Glycerophosphoinositol (GroPI) is the building block for phosphoinositide polyphosphates and is a substrate for PI-PLC. This item is non returnable