Learn More
Grainger Manufacturer Varies Ground Clamp, Steel, 500 A, 1/0 to 3/0 AWG, 1 ea
Catalog No. 502779235
Encompass_Preferred
This ground clamp provides a reliable electrical connection for welding applications, featuring a durable steel body with a braided copper shunt for effective conductivity. It is designed for use with cables ranging from 1/0 AWG to 3/0 AWG and is capable of accommodating various surface types including round pipes and flat plates.
  • 500 A maximum current capacity
  • High-tensile spring ensures a tight grip
  • 2 in. maximum jaw opening
  • 3 3/8 in. throat depth
  • Compatible with flat, pipe, and round surfaces
  • Features non-conductive components
Catalog No. 50-277-9235 Supplier Grainger Supplier No. 19N766
This ground clamp provides a reliable electrical connection for welding applications, featuring a durable steel body with a braided copper shunt for effective conductivity. It is designed for use with cables ranging from 1/0 AWG to 3/0 AWG and is capable of accommodating various surface types including round pipes and flat plates.
  • 500 A maximum current capacity
  • High-tensile spring ensures a tight grip
  • 2 in. maximum jaw opening
  • 3 3/8 in. throat depth
  • Compatible with flat, pipe, and round surfaces
  • Features non-conductive components