Learn More
Grainger Zoro Select C-Style Ground Clamp, 700 A, Brass, 1/0 AWG to 3/0 AWG, 1 ea
Catalog No. 502779212
Encompass_Preferred
This C-style ground clamp is designed for secure welding and industrial grounding applications, providing reliable connectivity for cables ranging from 1/0 AWG to 3/0 AWG. Featuring a durable brass body and zinc-plated steel screw, it offers excellent corrosion resistance and electrical conductivity for demanding environments. The clamp utilizes an integrated hand screw for manual tension adjustment, ensuring a firm grip on flat plates, pipes, and other round or flat surfaces.
  • 700 A maximum current capacity
  • 1/0 AWG to 3/0 AWG cable compatibility
  • 2.5 in. (6.35 cm) maximum jaw opening
  • 2 in. (5.08 cm) throat depth
  • Constructed with a brass body and zinc-plated steel screw
  • Suitable for flat, pipe, and round surface applications
Catalog No. 50-277-9212 Supplier Grainger Supplier No. 19N771
This C-style ground clamp is designed for secure welding and industrial grounding applications, providing reliable connectivity for cables ranging from 1/0 AWG to 3/0 AWG. Featuring a durable brass body and zinc-plated steel screw, it offers excellent corrosion resistance and electrical conductivity for demanding environments. The clamp utilizes an integrated hand screw for manual tension adjustment, ensuring a firm grip on flat plates, pipes, and other round or flat surfaces.
  • 700 A maximum current capacity
  • 1/0 AWG to 3/0 AWG cable compatibility
  • 2.5 in. (6.35 cm) maximum jaw opening
  • 2 in. (5.08 cm) throat depth
  • Constructed with a brass body and zinc-plated steel screw
  • Suitable for flat, pipe, and round surface applications