Learn More
Grainger Approved Vendor Ground Clamp, Steel, 200 A, 1.5 in. Jaw, 1 ea.
Catalog No. 502783429
Encompass_Preferred
This ground clamp is designed for secure electrical grounding applications, featuring a durable steel construction with a braided copper shunt for reliable conductivity. It is engineered to handle up to 200 A and is compatible with cable gauges ranging from 5 AWG to 3 AWG. This clamp provides a secure connection to flat, pipe, and round surfaces.
  • High-tensile spring provides a firm, tight grip
  • Compatible with flat, flat plate, pipe, and round surfaces
  • Supports cable gauges from 5 AWG to 3 AWG
  • Durable steel body construction with braided copper shunt
  • Maximum jaw opening of 1 1/2 in.
Catalog No. 50-278-3429 Supplier Grainger Supplier No. 19N765
This ground clamp is designed for secure electrical grounding applications, featuring a durable steel construction with a braided copper shunt for reliable conductivity. It is engineered to handle up to 200 A and is compatible with cable gauges ranging from 5 AWG to 3 AWG. This clamp provides a secure connection to flat, pipe, and round surfaces.
  • High-tensile spring provides a firm, tight grip
  • Compatible with flat, flat plate, pipe, and round surfaces
  • Supports cable gauges from 5 AWG to 3 AWG
  • Durable steel body construction with braided copper shunt
  • Maximum jaw opening of 1 1/2 in.