Learn More
Grainger Approved Vendor Ground Clamp, 600 A, 1/0 AWG to 3/0 AWG, Brass
Catalog No. 502779532
Encompass_Preferred
These flat-jaw ground clamps are designed for secure grounding on a variety of surfaces, including flat bar stock, metal plates, pipes, and round materials. Featuring a robust brass body and a high-tensile spring, the clamp ensures consistent pressure and reliable connectivity during arc welding operations.
  • 600 A maximum current capacity
  • Compatible with 1/0 AWG to 3/0 AWG cable
  • Features a ball point screw connection
  • High-tensile spring mechanism for secure gripping
  • Durable brass construction
  • Maximum jaw opening of 1-1/2 in.
  • Compatible with flat surfaces, plates, and pipes
Catalog No. 50-277-9532 Supplier Grainger Supplier No. 19N769
These flat-jaw ground clamps are designed for secure grounding on a variety of surfaces, including flat bar stock, metal plates, pipes, and round materials. Featuring a robust brass body and a high-tensile spring, the clamp ensures consistent pressure and reliable connectivity during arc welding operations.
  • 600 A maximum current capacity
  • Compatible with 1/0 AWG to 3/0 AWG cable
  • Features a ball point screw connection
  • High-tensile spring mechanism for secure gripping
  • Durable brass construction
  • Maximum jaw opening of 1-1/2 in.
  • Compatible with flat surfaces, plates, and pipes