Learn More
Grainger Approved Vendor Ground Clamp, 300 A, 3 AWG to 1 AWG, Brass, 1 ea
Catalog No. 502782893
Encompass_Preferred
This flat-jaw ground clamp is designed for secure electrical grounding on diverse surfaces including flat plates, pipes, and round stock. Featuring a durable brass body and a high-tensile spring, it ensures a tight, reliable grip during arc welding applications.
  • Brass construction for efficient conductivity
  • High-tensile spring mechanism to maintain firm jaw pressure
  • Compatible with cable sizes from 3 AWG to 1 AWG
  • Supports up to 300 A maximum current
  • Jaw opening capacity of 1 in.
  • Throat depth of 3-3/8 in.
Catalog No. 50-278-2893 Supplier Grainger Supplier No. 19N767
This flat-jaw ground clamp is designed for secure electrical grounding on diverse surfaces including flat plates, pipes, and round stock. Featuring a durable brass body and a high-tensile spring, it ensures a tight, reliable grip during arc welding applications.
  • Brass construction for efficient conductivity
  • High-tensile spring mechanism to maintain firm jaw pressure
  • Compatible with cable sizes from 3 AWG to 1 AWG
  • Supports up to 300 A maximum current
  • Jaw opening capacity of 1 in.
  • Throat depth of 3-3/8 in.