Learn More
Grainger Florock Ground Silica SL, Crystalline Silica, White, 50 lb
Catalog No. 502779605
Encompass_Preferred
Ground Silica SL is a crystalline silica and quartz filler designed to create skid-resistant surfaces when incorporated into coatings. This additive is compatible with epoxy, latex, and oil-based paints, stains, or sealers, making it suitable for enhancing safety on concrete, wood, metal, and fiberglass substrates in interior environments.
  • Provides skid-resistant properties for industrial stairs, decks, and walkways.
  • Compatible with resins to create thickened pastes or mortars for patching, filling holes and cracks, and vertical applications.
  • Facilitates the creation of self-leveling floor properties.
  • Supplied in a 50 lb container.
  • Meets multiple safety and quality standards, including ANSI, WHMIS, and OSHA requirements.
Catalog No. 50-277-9605 Supplier Grainger Supplier No. 19NZ17
Ground Silica SL is a crystalline silica and quartz filler designed to create skid-resistant surfaces when incorporated into coatings. This additive is compatible with epoxy, latex, and oil-based paints, stains, or sealers, making it suitable for enhancing safety on concrete, wood, metal, and fiberglass substrates in interior environments.
  • Provides skid-resistant properties for industrial stairs, decks, and walkways.
  • Compatible with resins to create thickened pastes or mortars for patching, filling holes and cracks, and vertical applications.
  • Facilitates the creation of self-leveling floor properties.
  • Supplied in a 50 lb container.
  • Meets multiple safety and quality standards, including ANSI, WHMIS, and OSHA requirements.