Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NewPath Learning™ Group 1 Elements 3D Model Making Kit
SCI_ED

Catalog No. S436054 Shop All NewPath Learning Products
Change view
Click to view available options
Grade:
6 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S436054 6 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S436054 Supplier NewPath Learning™ Supplier No. 247911
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The Group 1 Elements 3D Model Making Kit allows students to build models of atoms and electrons to see how alkali metals react and how bonds are formed.

  • Group 1 of the Periodic Table contains the reactive alkali metals.
  • Your students will first build their easy to make paper models of atoms and electrons.
  • They will then use the models to look at how the Group 1 elements react.
  • The differently colored electrons make it easy to see how the bonds are formed and the models shows reactions with water, oxygen, and chlorine.
  • The Model Making Kit includes enough materials for students to assemble up to 5 paper models working cooperatively.
  • The comprehensive Teacher Resource Guide included provides illustrated readings, activities, key vocabulary and an assessment, along with teaching opportunities for students to Engage, Explore, Explain, Extend and Evaluate their understanding of the concepts presented.

Specifications

For Use With (Application) Model Assembly
Grade 6 to 10
Includes 5 Models, Teacher Resource Guide, Vinyl Pouch
Length (English) 12 in.
Material Paper
Weight (English) 2 lb.
Width (English) 9 in.
Product Type 3D Model
Grade Level Intermediate - Middle School, High School
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.