Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ GROUP A STREPTOCOCCUS (GAS) CONTROL PANEL
SDP

Catalog No. 23047304 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
12 Swabs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-304 12 Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-304 Supplier Microbiologics™ Supplier No. 8219
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Group A Streptococcus (GAS) Control Panel includes 6 positive control swabs, 6 negative control swabs, and a Certificate of Analysis.

  • Shipping restriction: Currently not avaliable for shipment to Canada and Europe
  • Test Method: Molecular Testing

Specifications

Biosafety Level 1
Content And Storage 2°C to 8°C
Format Helix Elite
Manufacturing Origin United States
Product Type Helix Elite Control Reference Panels and Standards
For Use With (Application) For in vitro diagnostic use
Packaging Type Plastic Box
Quantity 12 Swabs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.