Learn More
Electron Microscopy Sciences Group Of 6-Variety Wash Bottle, 500 ml
SDP
Catalog No. 5027787
Encompass_Preferred
Permanently Labeled/Color-Coded Wash Bottles. These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz (500 ml), low density polyethylene. Includes Acetone, Menthanol, Water, Iso-propyl Alcohol, Ethanol.
Volume: 500 ml
Catalog No. 50-277-87 Supplier Electron Microscopy Sciences Supplier No. 64167
Permanently Labeled/Color-Coded Wash Bottles. These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz (500 ml), low density polyethylene. Includes Acetone, Menthanol, Water, Iso-propyl Alcohol, Ethanol.
Volume: 500 ml