Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC GROUT CLEANER

Catalog No. 502819345
Encompass

ZEP Grout Cleaner & Brightener is a highly acidic liquid cleaner designed to effectively remove tough stains and discoloration from grout. Its powerful formula is suitable for professional cleaning applications.

  • Removes tough stains and discoloration
  • Liquid form
  • Characteristic scent
  • Density of 1.06 g/cm³
  • Ph less than 1
  • Soluble in water
  • Formulated with urea hydrochloride and hydrochloric acid

Catalog No. 50-281-9345 Supplier MSC Supplier No. 17595141
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ZEP Grout Cleaner & Brightener is a highly acidic liquid cleaner designed to effectively remove tough stains and discoloration from grout. Its powerful formula is suitable for professional cleaning applications.

  • Removes tough stains and discoloration
  • Liquid form
  • Characteristic scent
  • Density of 1.06 g/cm³
  • Ph less than 1
  • Soluble in water
  • Formulated with urea hydrochloride and hydrochloric acid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.