Learn More
UPM KYMMENE INC GROWDEX -T READY USE HYDROGEL
Catalog No. NC1864934
Encompass
GrowDex®-T - ready to use hydrogel with superior imaging properties.GrowDex®-T is a ready to use hydrogel that has been specifically developed to offer superior imaging properties, but without compromising on any of the advantages of our original hydrogel GrowDex. GrowDex-T can be mixed directly with cells and culture media, no gelation or cross-linking step required. Ambient handling and shear thinning properties of GrowDex-T enable easy dispensing and use in automated systems.
Catalog No. NC1864934 Supplier UPM KYMMENE INC Supplier No. 200103002
GrowDex®-T - ready to use hydrogel with superior imaging properties.GrowDex®-T is a ready to use hydrogel that has been specifically developed to offer superior imaging properties, but without compromising on any of the advantages of our original hydrogel GrowDex. GrowDex-T can be mixed directly with cells and culture media, no gelation or cross-linking step required. Ambient handling and shear thinning properties of GrowDex-T enable easy dispensing and use in automated systems.