Learn More
Creative Biomart Growth Differentiation Factor 5 (GDF-5) Protein
Catalog No. 502084308
Encompass_Preferred
Growth Differentiation Factor 5 Human Recombinant produced in E.Coli is a homodimer, non-glycosylated polypeptide chain containing 2 x 120 amino acids and having a total molecular mass of 27.4kDa. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys. GDF5 is purified by proprietary chromatographic techniques.
Catalog No. 50-208-4308 Supplier Creative Biomart Supplier No. THP0257
Growth Differentiation Factor 5 Human Recombinant produced in E.Coli is a homodimer, non-glycosylated polypeptide chain containing 2 x 120 amino acids and having a total molecular mass of 27.4kDa. To enable bacterial expression the N-terminal sequence of Ala-Pro-Leu-Thr was replaced with a Lys. GDF5 is purified by proprietary chromatographic techniques.