Learn More
CPC Scientific Glp-Val-Asp-Ser-Met-Trp-Ala-Glu-Gln-Lys-Gln-Met-Glu-Leu-Glu-Ser-Ile-Leu-Val-Ala-Leu-Leu-Gln-Lys-His-Ser-Arg-Asn-Ser-Gln-Gly-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific GHRF003A
Encompass
ONE-LETTER SEQUENCE: Glp-VDSMWAEQKQMELESILVALLQKHSRNSQGMOLECULAR FORMULA: C153H248N44O50S2MOLECULAR WEIGHT:3568.1STORAGE CONDITIONS: -20 5CRESEARCH AREA: Hormonal
Catalog No. 50-210-9337