Learn More
BPS Bioscience Inc Growth Medium 1A, 500 ml, Liquid, 10% FBS, Sterile filtered, Mycoplasma-free, Low endotoxin
SDP
Catalog No. NC2102725
Encompass
The BPS Bioscience Growth Medium 1A is meticulously crafted to optimize the culturing of specific BPS Bioscience cell lines. It is a sterile-filtered liquid medium that supports cell proliferation and maintains cell line stability with the inclusion of selective antibiotics. Rigorous testing is performed on each batch to ensure functionality in recombinant cell growth while providing a mycoplasma-free and low-endotoxin environment. This growth medium is particularly suited for sensitive cell culture applications.
  • Optimized for specific BPS Bioscience cell lines.
  • Contains antibiotics for selective pressure on stable recombinant cell lines.
  • Ensures high purity, being sterile and low in endotoxin.
  • 10% FBS concentration for balanced nutrition.
  • Tested for growth of recombinant cells, guaranteeing reliability.
  • Stored in dark at 4°C with a shelf life of 6 months when thawed.
  • Available in a 500 ml liquid format.
This item is not returnable. View return policy
Catalog No. NC2102725 Supplier BPS Bioscience Inc Supplier No. 795281
Add to Cart
Add to Cart
The BPS Bioscience Growth Medium 1A is meticulously crafted to optimize the culturing of specific BPS Bioscience cell lines. It is a sterile-filtered liquid medium that supports cell proliferation and maintains cell line stability with the inclusion of selective antibiotics. Rigorous testing is performed on each batch to ensure functionality in recombinant cell growth while providing a mycoplasma-free and low-endotoxin environment. This growth medium is particularly suited for sensitive cell culture applications.
  • Optimized for specific BPS Bioscience cell lines.
  • Contains antibiotics for selective pressure on stable recombinant cell lines.
  • Ensures high purity, being sterile and low in endotoxin.
  • 10% FBS concentration for balanced nutrition.
  • Tested for growth of recombinant cells, guaranteeing reliability.
  • Stored in dark at 4°C with a shelf life of 6 months when thawed.
  • Available in a 500 ml liquid format.