Learn More
BPS Bioscience Inc Growth Medium 1C, 3 x 500 ml, This liquid Growth Medium is a complete medium optimized to culture select BPS Bioscience cell lines.
SDP
Catalog No. NC2823496
Encompass
Growth Medium 1C (BPS #79532): MEM medium supplemented with 10% FBS, 1% non-essential amino acids, 1 mM Na pyruvate, 1% Penicillin/Streptomycin plus 50 µg/ml of Hygromycin B
Catalog No. NC2823496 Supplier BPS Bioscience Inc Supplier No. 795322
Growth Medium 1C (BPS #79532): MEM medium supplemented with 10% FBS, 1% non-essential amino acids, 1 mM Na pyruvate, 1% Penicillin/Streptomycin plus 50 µg/ml of Hygromycin B