Learn More
BPS Bioscience Inc Growth Medium 1M, 1 L volume, Liquid Growth Medium, 10% FBS, 1% Non-essential Amino Acids, 1 mM Sodium Pyruvate, 1% Penicillin/Streptomycin, Mycoplasma-free
SDP
Catalog No. NC3104273
Encompass
Growth Medium 1M, 1 L volume, Liquid Growth Medium, 10% FBS, 1% Non-essential Amino Acids, 1 mM Sodium Pyruvate, 1% Penicillin/Streptomycin, Mycoplasma-free
Catalog No. NC3104273 Supplier BPS Bioscience Inc Supplier No. 79723
Growth Medium 1M, 1 L volume, Liquid Growth Medium, 10% FBS, 1% Non-essential Amino Acids, 1 mM Sodium Pyruvate, 1% Penicillin/Streptomycin, Mycoplasma-free