Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BPS Bioscience Inc Growth Medium 2A, 3 x 500 ml, Sterile-filtered, Mycoplasma-free, RPMI 1640 medium, 10% FBS, Selective antibiotics
SDP

Catalog No. NC2739153
Encompass

Growth Medium 2A is a sterile-filtered, mycoplasma-free liquid medium designed for the optimized culture of select cell lines. This complete growth medium contains RPMI 1640 supplemented with essential components to ensure effective maintenance of stable cell lines. Each batch undergoes rigorous quality control, confirming its low endotoxin level and compliance with research standards.

  • Complete liquid growth medium for cell culture
  • Formulated from RPMI 1640 medium (ATCC modification)
  • Contains 10% FBS and 1% Penicillin/Streptomycin
  • Includes selective antibiotics: 1 mg/ml Geneticin, 200 μg/ml Hygromycin B
  • Sterile filtered through 0.2 μm
  • Mycoplasma-free assurance
  • Low endotoxin level for enhanced safety
  • ISIA-certified FBS sourced from the USA
  • Stable for 6 months when stored at 4°C in the dark
  • Packaged in size of 3 x 500 ml
  • Designed for research use only

Catalog No. NC2739153 Supplier BPS Bioscience Inc Supplier No. 601902
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Growth Medium 2A is a sterile-filtered, mycoplasma-free liquid medium designed for the optimized culture of select cell lines. This complete growth medium contains RPMI 1640 supplemented with essential components to ensure effective maintenance of stable cell lines. Each batch undergoes rigorous quality control, confirming its low endotoxin level and compliance with research standards.

  • Complete liquid growth medium for cell culture
  • Formulated from RPMI 1640 medium (ATCC modification)
  • Contains 10% FBS and 1% Penicillin/Streptomycin
  • Includes selective antibiotics: 1 mg/ml Geneticin, 200 μg/ml Hygromycin B
  • Sterile filtered through 0.2 μm
  • Mycoplasma-free assurance
  • Low endotoxin level for enhanced safety
  • ISIA-certified FBS sourced from the USA
  • Stable for 6 months when stored at 4°C in the dark
  • Packaged in size of 3 x 500 ml
  • Designed for research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.