Learn More
BPS Bioscience Inc Growth Medium 2B, Liquid, 500 ml, RPMI 1640, 10% FBS, 1% Penicillin/Streptomycin, 1 mg/ml Geneticin, ISIA-certified, Sterile filtered, Mycoplasma-free
SDP
Catalog No. NC2282204
Encompass
Growth Medium 2B, Liquid, 500 ml, RPMI 1640, 10% FBS, 1% Penicillin/Streptomycin, 1 mg/ml Geneticin, ISIA-certified, Sterile filtered, Mycoplasma-free
This item is not returnable. View return policy
Catalog No. NC2282204 Supplier BPS Bioscience Inc Supplier No. 795301
Growth Medium 2B, Liquid, 500 ml, RPMI 1640, 10% FBS, 1% Penicillin/Streptomycin, 1 mg/ml Geneticin, ISIA-certified, Sterile filtered, Mycoplasma-free