Learn More
ACROBiosystems ACROBiosystems Rhesus macaque Growth Hormone R (GHR) Protein, Fc Tag
Catalog No. 501957040
Encompass_Preferred
Recombinant Protein;100UG;Rhesus macaque GHR, Fc Tag (GHR-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 264 (Accession # P79194-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.7 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GH, Tag Free at 2 μg/mL (100 μL/well) can bind Rhesus macaque GHR, Fc Tag (Cat. No. GHR-C5253) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-195-7040 Supplier ACROBiosystems Supplier No. GHRC5253100UG
Recombinant Protein;100UG;Rhesus macaque GHR, Fc Tag (GHR-C5253) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 264 (Accession # P79194-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.7 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human GH, Tag Free at 2 μg/mL (100 μL/well) can bind Rhesus macaque GHR, Fc Tag (Cat. No. GHR-C5253) with a linear range of 0.039-0.625 μg/mL (QC tested).