Learn More
ACROBiosystems Recombinant Protein;Human Growth Hormone R (GHR) Protein, Fc Tag;HEK293;50UG;GHR,GHR,GHBP,GH receptor,Growth Hormone R
Supplier:  ACROBiosystems GHRH525750UG
Encompass_Preferred
Recombinant Protein;Human Growth Hormone R (GHR) Protein, Fc Tag;50UG;Human GHR, Fc Tag (GHR-H5257) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Tyr 264 (Accession # P10912).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 54.3 kDa. As a result of glycosylation, the protein migrates as 66-85 kDa under reducing (R) condition, and 135-160 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human GH, Tag Free (Cat. No. GH1-H4117) at 2μg/mL (100 μL/well) can bind Human GHR, Fc Tag (Cat. No. GHR-H5257) with a linear range of 0.19-3.1 ng/mL (QC tested).
Catalog No. 50-165-8170