Learn More
Bachem GRP (14-27) (human, porcine, canine) Trifluoroacetate
SDP
Supplier:  Bachem 4011671.0005
Encompass_Preferred
GRP (14-27) (human, porcine, canine) Trifluoroacetate, H-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2, 81608-29-9, C75H110N24O16S2, Mr 1667.98, 5mg. Gastrin Releasing Peptide (14-27), porcine, human.
Catalog No. 50-194-7477