Learn More
Medchemexpress LLC HY-133735A 5mg Medchemexpress, GRP-60367 (hydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY133735A5MG
Encompass_Preferred
Medchemexpress, HY-133735A 5mg GRP-60367 (hydrochloride) CAS: GRP-60367 hydrochloride is a first-in-class small-molecule rabies virus (RABV) entry inhibitor with nanomolar potency against some RABV strains. GRP-60367 hydrochloride specifically targets the RABV G protein. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1347