Learn More
Enzo Life Sciences Grp 94 polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADISPA851F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of mouse Grp94. UniProt ID: P08113. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Bovine. Applications: ICC, IHC (PS), IP, WB. Recommended Dilutions: Immunoprecipitation (1:80) Western Blot (1:1,000) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7567