Learn More
Enzo Life Sciences Grp 94 polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPA851D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the C-terminus of mouse Grp94. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Bovine. Applications: ICC, IHC (PS), IP, WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7566