Learn More
Aviva Systems Biology Corp GRP Antibody - C-terminal region (ARP59548_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59548_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRP. It was validated on Western Blot. . Alternate names: BN, GRP-10, preproGRP, proGRPApplicationWB. Species reactivity: Human
Catalog No. 50-286-737