Learn More
Echelon Biosciences Research Labs Gastrin Releasing Peptide, human / GRP, h - 0.5 mg, CAS#: 93755-85-2
SDP
Catalog No. 501908515
Encompass_Preferred
Gastrin Releasing Peptide, human / GRP, h - 0.5mg , Val-Pro-Leu-Pro-Ala-Gly-Gly-Gly-Thr-Val-Leu-Thr-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-190-8515 Supplier Echelon Biosciences Research Labs Supplier No. 471850.5MG
Gastrin Releasing Peptide, human / GRP, h - 0.5mg , Val-Pro-Leu-Pro-Ala-Gly-Gly-Gly-Thr-Val-Leu-Thr-Lys-Met-Tyr-Pro-Arg-Gly-Asn-His-Trp-Ala-Val-Gly-His-Leu-Met-NH2 Category/Application: Neuropeptides Hormones This item is non returnable