Learn More
Enzo Life Sciences Gastrin releasing peptide polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences BMLGA11660100
Encompass_Preferred
Immunogen: Synthetic GRP. Source: Whole rabbit serum. Species reactivity: Species independent. Specificity: Recognizes GRP in a wide range of species. Applications: IHC (FS). Formulation: Liquid. Neat serum.
Catalog No. 50-201-0892