Learn More
Echelon Biosciences Research Labs GRP1-PH DOMAIN GST 5 ug
SDP
Supplier:  Echelon Biosciences Research Labs G39015UG
Encompass
Each vial of PI(3,4,5)P3 Grip contains lyophilized, recombinant GST-tagged GRP1-PH domain. This PI(3,4,5)P3-specific binding protein can be used as a control reagent with PIP Strips, PIP Arrays, and PIP Plates. It is easily reconstituted in water or buffered solutions. Each 2.5 g vial contains enough protein for use on one PIP Strip or PIP Array. PI(3,4,5)P3 Grip can be detected with commercial anti-GST antibodies. This item is non returnable
Catalog No. 50-720-349