Learn More
Medchemexpress LLC GRP75-IN-1 dihydrochloride | 2922680-16-6 | 99.8% | 539.60 | 50 MG
SDP
Catalog No. 5000392758
Encompass_Preferred
GRP75-IN-1 dihydrochloride is an anti-endometrial cancer (EC) agent that induces apoptosis. It reduces Ca2+ levels in mitochondria by targeting GRP75 and disrupting its interaction with IP3R.
  • Anti-endometrial cancer (EC) agent
  • Induces apoptosis
  • Reduces mitochondrial Ca2+ levels by targeting GRP75
  • Disrupts GRP75 interaction with IP3R
Catalog No. 50-003-92758 Supplier Medchemexpress LLC Supplier No. HY162774A50MG
GRP75-IN-1 dihydrochloride is an anti-endometrial cancer (EC) agent that induces apoptosis. It reduces Ca2+ levels in mitochondria by targeting GRP75 and disrupting its interaction with IP3R.
  • Anti-endometrial cancer (EC) agent
  • Induces apoptosis
  • Reduces mitochondrial Ca2+ levels by targeting GRP75
  • Disrupts GRP75 interaction with IP3R