Learn More
Enzo Life Sciences Grp75 Mortalin (human), (recombinant) (50µg)
SDP
Supplier:  Enzo Life Sciences ADISPP828D
Encompass_Preferred
Alternative name: MtHsp70, HspA9, Heat shock 70 kDa protein 9. Purity: ≥80% (SDS-PAGE; Western blot). MW: ~75kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 3% glycerol and 150mM sodium chloride. Source: Produced in E. coli. Long Term Storage: -80°C. Applications: WB .
Catalog No. 50-200-7647