Learn More
Enzo Life Sciences Grp75 Mortalin monoclonal antibody (30A5) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS7480200
Encompass_Preferred
Clone: 30A5. Isotype: IgG1κ. Immunogen: Native human Grp75 (Mortalin). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, Chicken, Bovine. Applications: IHC, WB. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3289