Learn More
Enzo Life Sciences Grp75 Mortalin monoclonal antibody (30A5) (50µl)
SDP
Supplier:  Enzo Life Sciences ADISPS825D
Encompass_Preferred
Clone: 30A5. Host: Mouse. Isotype: IgG1. Immunogen: Native human Grp75 (Mortalin). UniProt ID: P38646. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Drosophila, Dog, Chicken, Bovine. Applications: IHC, WB. Recommended Dilutions: Immunohistochemistry (1:50) Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Ammonium sulfate precipitation purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7660